missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GJA10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10313093
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10313093 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10313093 Supplier Invitrogen™ Supplier No. 405000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

40-5000 was used in immunofluorescence analysis of Connexin57 (Cx57) on sections of adult mouse retina.

Connexins, such as GJA10, are involved in the formation of gap junctions, intercellular conduits that directly connect the cytoplasms of contacting cells. Each gap junction channel is formed by docking of 2 hemichannels, each of which contains 6 connexin subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GJA10
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJA10
Gene Accession No. Q969M2, Q9WUS4
Gene Alias connexin 57; connexin 62; connexin-57; Connexin-62; cx57; Cx-57; Cx59; CX62; gap junction alpha-10 protein; gap junction membrane channel protein alpha 10; gap junction protein alpha 10; gap junction protein connexin 57; gap junction protein, alpha 10; gap junction protein, alpha 10, 62kDa; GJA10; LOW QUALITY PROTEIN: gap junction alpha-10 protein; RGD1309630
Gene Symbols GJA10
Host Species Rabbit
Immunogen Synthetic peptide derived from an internal region of the mouse, rat, Chinese hamster and golden hamster connexin 57 (Cx57) proteins.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14610, 84694
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.