missing translation for 'onlineSavingsMsg'
Learn More

Glufosinate ammonium, 95%, Thermo Scientific™

Glufosinate ammonium, 95%, CAS # 77182-82-2, also known as glufosinate, is a non-selective phosphorous herbicide.

Brand:  Thermo Scientific Alfa Aesar J66186.03

Quantity 1g

 View more versions of this product

Product Code. 15407449

  • ONLINE EXCLUSIVE PRICE
    332.00€
    324.99€ / 1g
    Save 7.01€
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Glufosinate ammonium is a compound which exhibits broad-spectrum herbicidal properties through inhibition of glutamine synthetase, which disrupts cell membranes

Application

• Glufosinate ammonium acts as a non-selective herbicide

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

77182-82-2
198.16
ZBMRKNMTMPPMMK-UHFFFAOYNA-N
122130249
[NH4+].CP(O)(=O)CCC(N)C([O-])=O
C5H15N2O4P
MFCD00055562,MFCD00211362
DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt
ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate

Specifications

77182-82-2
1g
C5H15N2O4P
8163399
14,7339
Soluble in water
[NH4+].CP(O)(=O)CCC(N)C([O-])=O
198.16
198.16
Glufosinate ammonium
White
>110°C (230°F)
MFCD00055562,MFCD00211362
519°C
DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt
ZBMRKNMTMPPMMK-UHFFFAOYNA-N
ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate
122130249
95%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.