missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GluR1 Polyclonal Antibody

Product Code. 16508791
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16508791 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16508791 Supplier Invitrogen™ Supplier No. PA595946

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PWGQGIDIQR ALQQVRFEGL TGNVQFNEKG RRTNYTLHVI EMKHDGIRKI GYWNEDDKFV PAATDAQAGG DNSSVQNRTY IVTTILEDPY VMLKKNANQF EGNDRYEGYC VELAAEIAKH VGYSYRLEIV SDGKYGARDP D Positive Samples: BT-474; Cellular Location: Cell junction, Cell membrane, Cell projection, Endoplasmic reticulum membrane, Multi-pass membrane protein, dendrite, dendritic spine, postsynaptic cell membrane, postsynaptic density, synapse.

GluR1 (glutamate receptor 1) is an ionotropic glutamate receptor. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter, L-glutamate, induces a conformation change that leads to the opening of the cation channel and converts the chemical signal to an electrical impulse. Then, the receptor desensitizes rapidly and enters a transiet inactive state characterized by the presence of a bound agonist. In the presence of CACNG4, CACNG7, or CACNG8 GluR1 shows resensitization which is characterized by a delayed accumulation of current flux upon continued application of glutamate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GRIA1
Gene Accession No. P19490, P23818, P42261
Gene Alias 2900051M01Rik; AI853806; AMPA 1; AMPA glutamate receptor; AMPA receptor GluR1/A; AMPA-selective glutamate receptor 1; Glr1; Glr-1; GluA1; GLUH 1; GLUH1; GluR K1; GLUR1; Glur-1; GluR1 flip; GluR1 flop; GLURA; GluR-A; GluRK1; gluR-K1; glutamate ionotropic receptor AMPA type subunit 1; glutamate receptor; glutamate receptor 1; glutamate receptor A; glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA1 (alpha 1); GPRC1A; Gria 1; Gria1; GRM1A; HBGR1; HGBR1; HIPA; HIPA1; MGC13325; MGC133252; NMDA1; NMDAR1; NR1; OTTHUMP00000160643; OTTHUMP00000165781; OTTHUMP00000224241; OTTHUMP00000224242; OTTHUMP00000224243; RP11-350O14.1
Gene Symbols GRIA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 330-470 of human GluR1/GRIA1 (NP_0008182).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14799, 2890, 50592
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.