missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GluR3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total GRIA3.
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties.
Specifications
Specifications
| Antigen | GluR3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GRIA3 |
| Gene Accession No. | P19492, P42263, Q9Z2W9 |
| Gene Alias | 2900064I19Rik; AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA3; GLUR3; Glur-3; Gluralpha3; GLURC; GLUR-C; GluR-K3; glutamate ionotropic receptor AMPA type subunit 3; glutamate receptor; glutamate receptor 3; glutamate receptor C; glutamate receptor channel alpha3 subunit; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA3 (alpha 3); GRIA3; Kiaa4184; LOW QUALITY PROTEIN: glutamate receptor 3; MRX94; OTTHUMP00000024262; RP11-349N19.3 |
| Gene Symbols | GRIA3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?