Learn More
Glutamine Synthetase Mouse anti-Human, Unconjugated, Clone: 1B6G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66323-2-IG-150UL
Description
The Glutamine Synthetase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glutamine Synthetase. This antibody recognizes human, mouse, rat antigen. The Glutamine Synthetase antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Glutamine Synthetase | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Immunohistochemistry, ELISA | |
| 1B6G4 | |
| Unconjugated | |
| 150 μL | |
| 2752 | |
| IgG1 |