Learn More
Glutamine Synthetase Mouse anti-Human, Unconjugated, Clone: 1B6G4, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Glutamine Synthetase |
|---|---|
| Clone | 1B6G4 |
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16838095
|
Proteintech
66323-2-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16828095
|
Proteintech
66323-2-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Glutamine Synthetase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glutamine Synthetase. This antibody recognizes human, mouse, rat antigen. The Glutamine Synthetase antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Glutamine Synthetase | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 2752 | |
| Primary |
| 1B6G4 | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.