Learn More
Glutamine Synthetase Mouse anti-Human, Unconjugated, Clone: 1B6G4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66323-2-IG-20UL
 View more versions of this product
Product Code. 16838095
  • 145.00€ / 20µL
Description
The Glutamine Synthetase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glutamine Synthetase. This antibody recognizes human, mouse, rat antigen. The Glutamine Synthetase antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
Glutamine Synthetase
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Immunohistochemistry, ELISA
1B6G4
Unconjugated
20 μL
2752