missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde (50% in water), TRC

Product Code. 30012801
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012801 10 g 10g
30009169 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012801 Supplier Toronto Research Chemicals Supplier No. TRCG59730010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5 H8 O2
Molecular Weight (g/mol) 100.12
Synonym Glutaraldehyde (6CI,8CI), 1,5-Pentanedial, 5-Oxopentanal, Aldesan, Aqucar 550, Bactron K 31, Banicide, Bioban Eth 1000, Biocide G 30, Biomate 743, Cidex, Cidex 7, Cidex-Dialyzer, Cidexplus, Cleancide 275, Diglutaric aldehyde, Eimaldehyde, Floperm 665X1, Formula H, GT 50, Glu-Cid, Glutaclean, Glutaral, Glutardialdehyde, Glutarex 28, Glutaric acid dialdehyde, Glutaric dialdehyde, Glutohyde, Hospex, KS 02, Kcide 850, Maxicide Plus, Metricide Plus, NSC 13392, Odycide B 310, Panavirocide, Permachem OB 5, Piror 850, Protectol GA 50, Relugan GT, Relugan GT 50, Relugan GTW, Sellatan CF-N, Sonacide, Sporicidin, Sterihyde, Sterihyde L, Sterisol S, Surcide G 50, T 352, Tret-O-Lite XC 102, Ucarcide 250, Ucarcide 250 Antimicrobial, Ucarcide 50, Ucarcide 50 Antimicrobial, Ukatan 24, Venno-FF super, Wavicide 01
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2
Formula Weight 100.0524
Chemical Name or Material Glutaraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.