missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM130/GOLGA2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP2-53420
This item is not returnable.
View return policy
Description
GM130/GOLGA2 Polyclonal specifically detects GM130/GOLGA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GM130/GOLGA2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| GOLGA2 | |
| Partial recombinant human GM130/GOLGA2 protein (amino acids 528-606). [UniPro Q08379] | |
| 0.1 mg | |
| Cellular Markers, Golgi Apparatus Markers | |
| 2801 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.5 ug/ml, Immunohistochemistry 1:1000 - 1:1500, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:1500 | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction