missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM130/GOLGA2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
195.00€ - 508.00€
Specifications
| Antigen | GM130/GOLGA2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5 ug/ml, Immunohistochemistry 1:1000 - 1:1500, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:1500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18040324
|
Novus Biologicals
NBP2-53420SS |
0.025 mg |
195.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18050584
|
Novus Biologicals
NBP2-53420 |
0.1 mg |
508.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GM130/GOLGA2 Polyclonal specifically detects GM130/GOLGA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GM130/GOLGA2 | |
| Western Blot 0.5 ug/ml, Immunohistochemistry 1:1000 - 1:1500, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:1500 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Golgi Apparatus Markers | |
| PBS with 0.02% Sodium Azide | |
| 2801 | |
| Partial recombinant human GM130/GOLGA2 protein (amino acids 528-606). [UniPro Q08379] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| GOLGA2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title