Learn More
GMEB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13314-1-AP-20UL
 View more versions of this product
Product Code. 16881454
  • 145.00€ / 20µL
Description
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB40, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Single nucleotide polymorphisms near ZBTB40 have been linked to differences in bone mineral density of the hip and lumbar spine, and had a significant association with low-trauma fractures suggesting that ZBTB40 may be involved in bone development and formation.
Specifications
Specifications
GMEB2
Polyclonal
Unconjugated
GMEB2
DNA binding protein p79PIF, GMEB 2, GMEB2, KIAA1269, P79PIF, PIF p79, PIF79
Rabbit
Antigen Affinity Chromatography
RUO
229004, 26205, 83635
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88873, P58929, Q9UKD1
Gmeb2
GMEB2 Fusion Protein Ag4157
20 μL
Primary
Mouse, Human, Rat