missing translation for 'onlineSavingsMsg'
Learn More

GMEB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen GMEB2
Concentration 0.35 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16881454
View Documents
Proteintech
13314-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16871454
View Documents
Proteintech
13314-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB40, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Single nucleotide polymorphisms near ZBTB40 have been linked to differences in bone mineral density of the hip and lumbar spine, and had a significant association with low-trauma fractures suggesting that ZBTB40 may be involved in bone development and formation.
Specifications

Specifications

GMEB2
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
O88873, P58929, Q9UKD1
229004, 26205, 83635
GMEB2 Fusion Protein Ag4157
Primary
-20°C
GMEB2
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
DNA binding protein p79PIF, GMEB 2, GMEB2, KIAA1269, P79PIF, PIF p79, PIF79
Gmeb2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.