missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GOLT1A Polyclonal Antibody
GREENER_CHOICE

Product Code. 16673995
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16673995 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16673995 Supplier Invitrogen™ Supplier No. PA599751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GOLT1A.

GOLT1A may be involved in fusion of ER-derived transport vesicles with the Golgi complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLT1A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GOLT1A
Gene Accession No. Q6ZVE7, Q9DCQ3
Gene Alias 0610012C01Rik; CGI-141; Golgi transport 1 homolog A; golgi transport 1 homolog A (S. cerevisiae); golgi transport 1A; golt1a; GOT1; GOT1B; hGOT1b; HMFN1187; Kiss1; metastatin; putative protein product of HMFN1187; RGD1566310; RP11-203F10.3; Vesicle transport protein GOT1A; YMR292W
Gene Symbols GOLT1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human GOLT1A(Accession Q6ZVE7), corresponding to amino acid residues L34-L84.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127845, 68338
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.