missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPATCH8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15718521 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15718521 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15718521 Supplier Invitrogen™ Supplier No. PA560694

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SIASVFKDHA EEGTSEDGTK PDEKSSDQGL QKVGDSDGSS NLDGKKEDED PQDGGSLAST LSKLKRMKRE EGAGATEPEY YHYIPPAHCK VKPN Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

The GPATCH8 protein contains an RNA-processing domain, a zinc finger domain, a lysine-rich region and a serine-rich region. A mutation in the serine-rich region of the protein is thought to be associated with hyperuricemia (PMID: 21594610). Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPATCH8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPATCH8
Gene Accession No. Q9UKJ3
Gene Alias 5430405G24Rik; AU018890; ENSMUSG00000075516; Fbm1; Fyn-binding molecule 1; G patch domain containing 8; G patch domain-containing protein 8; Gpatc8; G-patch domain containing 8; GPATCH8; KIAA0553; mKIAA0553
Gene Symbols GPATCH8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPATCH8. Recombinant protein control fragment (Product #RP-93525).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.