missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR12 Polyclonal Antibody

Product Code. 15304511
Change view
Click to view available options
Quantity:
50 μg
Conditionnement:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
15304511 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15304511 Fournisseur Invitrogen™ Code fournisseur PA533617

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Mouse, Panda, Dog, Pig (94%).

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR12 is an orphan G-protein-coupled receptor which constitutively activates the Gs protein and increases intracellular cAMP. GPR12 can also be activated by sphingosylphosphorylcholine to promote various cellular functions which include proliferation, neurite outgrowth, cell survival and differentiation.
TRUSTED_SUSTAINABILITY

Spécification

Antigen GPR12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR12
Gene Accession No. P47775
Gene Alias FLJ18149; FLJ97704; G protein-coupled receptor 12; GPCR01; GPCR12; Gpcr20; Gpcr21; Gpr12; G-protein coupled receptor 12; MGC138349; MGC138351; PPP1R84; protein phosphatase 1, regulatory subunit 84; R334
Gene Symbols GPR12
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR12 / GPCR12.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2835
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.