missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR137 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
GPR137 is a protein coding gene.
Specifications
Specifications
| Antigen | GPR137 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | GPR137 |
| Gene Accession No. | Q96N19 |
| Gene Alias | AI428855; C11orf4; G protein-coupled receptor 137; G protein-coupled receptor TM7SF1L1; GPR137; GPR137A; integral membrane protein GPR137; TM7SF1L1; transmembrane 7 superfamily member 1-like 1 protein |
| Gene Symbols | GPR137 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?