missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR34 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15392611
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15392611 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15392611 Supplier Invitrogen™ Supplier No. PA533697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Bovine, Horse (100%) Marmoset, Mouse, Rat, Elephant, Guinea pig (94%).

G protein-coupled receptors (GPCRs), such as GPR34, are integral membrane proteins containing 7 putative transmembrane domains (TMs). These proteins mediate signals to the interior of the cell via activation of heterotrimeric G proteins that in turn activate various effector proteins, ultimately resulting in a physiologic response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR34
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Gpr34
Gene Accession No. Q9UPC5
Gene Alias G protein-coupled receptor 34; Gpr34; G-protein-coupled receptor; G-protein-coupled receptor GPR34; Lypsr1; probable G-protein coupled receptor 34
Gene Symbols Gpr34
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 2nd cytoplasmic domain of human GPR34.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050641, 100139092, 2857
Target Species Human, Bovine, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.