missing translation for 'onlineSavingsMsg'
Learn More

H-2Kd/H-2Dd Mouse anti-Mouse, BV 421, Clone: 34-1-2S, BD Horizon™

Product Code. 17611426
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17611426 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17611426 Supplier BD Biosciences Supplier No. 567294

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

H-2Kd/H-2Dd Monoclonal antibody specifically detects H-2Kd/H-2Dd in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2Kd/H-2Dd
Applications Flow Cytometry
Classification Monoclonal
Clone 34-1-2S
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias H2-D1, H-2D; H2-K1, H-2K, H2-K
Host Species Mouse
Immunogen C57BL/6 × DBA/2 (BDF1) splenocytes
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.