missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol, TRC

Product Code. 18060330
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18060330 1 g 1g
18020740 5 g 5g
18070630 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18060330 Supplier Toronto Research Chemicals Supplier No. TRCH1037001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-86-8
Molecular Formula C21 H23 Cl F N O2
Molecular Weight (g/mol) 375.86
Synonym 1-Butanone, 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-, Butyrophenone, 4-[4-(p-chlorophenyl)-4-hydroxypiperidino]-4'-fluoro- (6CI,8CI), 1-(3-p-Fluorobenzoylpropyl)-4-p-chlorophenyl-4-hydroxypiperidine, 4-(4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl)-1-(4-fluorophenyl)-1-butanone, 4-(4-Hydroxy-4'-chloro-4-phenylpiperidino)-4'-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4'-fluorobutyrophenone, Aloperidin, Bioperidolo, Brotopon, Dozic, Einalon S, Eukystol, Haldol, Haloperidol, Haloperin, Halosten, Keselan, Linton, McN-JR 1625, NSC 170973, NSC 615296, Neurodol, Peluces, R 1625, Serenace, Serenase, Serenelfi, Sigaperidol, γ-[4-(p-Chlorphenyl)-4-hydroxypiperidino]-p-fluorbutyrophenone
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
SMILES OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2
Formula Weight 375.1401
Percent Purity >95%
Chemical Name or Material Haloperidol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.