missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAND1 Antibody (8E7A11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-51547-0.025ml
This item is not returnable.
View return policy
Description
HAND1 Monoclonal specifically detects HAND1 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| HAND1 | |
| Monoclonal | |
| Unconjugated | |
| BHLHA27, bHLHa27heart- and neural crest derivatives-expressed protein 1, Class A basic helix-loop-helix protein 27, EHAND, Extraembryonic tissues, heart, autonomic nervous system and neural crestderivatives-expressed protein 1, heart and neural crest derivatives expressed 1, Hxt, Thing1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| 9421 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 8E7A11 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| HAND1 | |
| Purified recombinant fragment of HAND1 (aa90-190) expressed in E. Coli. | |
| 0.025 mL | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction