missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCFC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15720221 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
15720221 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15720221 Fournisseur Invitrogen™ Code fournisseur PA552267

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Immunogen sequence: APSQVQLIKA TTNSFHVKWD EVSTVEGYLL QLSTDLPYQA ASSDSSAAPN MQGVRMDPHR QGSNNIVPNS INDTINSTKT EQPATKETSM KNKPDFKALT DSNAILYPSL ASNASNHNSH VVDMLRKNEG PHTSANVGV Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 60%.

This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described.
TRUSTED_SUSTAINABILITY

Spécification

Antigen HCFC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HCFC2
Gene Accession No. Q9Y5Z7
Gene Alias 1700129L13Rik; AU019838; C2 factor; HCF2; HCF-2; HCFC2; Host cell factor 2; host cell factor C2
Gene Symbols HCFC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HCFC2. Recombinant protein control fragment (Product #RP-89327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.