missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NB100-1669SS
This item is not returnable.
View return policy
Description
HDAC3 Polyclonal specifically detects HDAC3 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| HDAC3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HDAC3 | |
| This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 2-17 of human HDAC3. | |
| 0.025 mg | |
| Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases | |
| 8841 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 2 - 5ug/ml, Simple Western 1:25, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 | |
| EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HeLa, a 50 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction