missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
231.00€ - 536.00€
Specifications
| Antigen | HDAC3 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 - 5ug/ml, Simple Western 1:25, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18061764
|
Novus Biologicals
NB100-1669SS |
0.025 mg |
231.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18757883
|
Novus Biologicals
NB100-1669 |
0.1 mg |
536.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC3 Polyclonal specifically detects HDAC3 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| HDAC3 | |
| Western Blot 2 - 5ug/ml, Simple Western 1:25, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
| HDAC3 | |
| IgG | |
| Protein G purified | |
| In HeLa, a 50 kDa band is observed. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases | |
| PBS with 0.05% Sodium Azide | |
| 8841 | |
| This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 2-17 of human HDAC3. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title