missing translation for 'onlineSavingsMsg'
Learn More

Hematein (Technical Grade), TRC

Product Code. 30004835
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004835 1 g 1g
30012560 2.5 g 2.5g
30007620 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004835 Supplier Toronto Research Chemicals Supplier No. TRCH2456531G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475-25-2
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.28
Synonym 6a,7-Dihydro-3,4,6a,10-tetrahydroxybenz[b]indeno[1,2-d]pyran-9(6H)-one, Haematein, NSC 7333
IUPAC Name 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one
SMILES OC1=CC2=C3c4ccc(O)c(O)c4OCC3(O)CC2=CC1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H12O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,18-21H,5-6H2
Formula Weight 300.0634
Chemical Name or Material Haematein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.