Learn More
Hemoglobin beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14081
This item is not returnable.
View return policy
Description
Hemoglobin beta Polyclonal specifically detects Hemoglobin beta in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| IgG |