missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 559.00€
Specifications
| Antigen | Hemoglobin beta |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18436071
|
Novus Biologicals
NBP2-14081-25ul |
25ul |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769483
|
Novus Biologicals
NBP2-14081 |
559.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Hemoglobin beta Polyclonal specifically detects Hemoglobin beta in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Hemoglobin beta | |
| Polyclonal | |
| Rabbit | |
| Human | |
| beta globin chain, beta-globin, CD113t-C, HBBB, HBD, Hemoglobin beta chain, hemoglobin subunit beta, hemoglobin, beta | |
| HBB | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 3043 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FATLSELHCDKLHVDPENFRLLGN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title