missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HEN2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16644025
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16644025 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16644025 Supplier Invitrogen™ Supplier No. PA599778

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HEN2.

May serve as DNA-binding protein and may be involved in the control of cell-type determination, possibly within the developing nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NHLH2
Gene Accession No. Q02577, Q64221
Gene Alias 6230401I09Rik; bHLHa34; bHLHa35; class A basic helix-loop-helix protein 34; Class A basic helix-loop-helix protein 35; helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); helix-loop-helix protein 2; HEN1; HEN-1; Hen2; HEN-2; KIAA0490; Nescient helix loop helix 1; nescient helix loop helix 2; nescient helix-loop-helix 1; nescient helix-loop-helix 2; Nhlh1; Nhlh2; Nscl; NSCL1; NSCL-1; NSCL2; NSCL-2; Tal2
Gene Symbols NHLH2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NHLH2(Accession Q02577), corresponding to amino acid residues A17-E67.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18072, 4808
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.