missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody (C7-50) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2740-0.025mg
This item is not returnable.
View return policy
Description
Hepatitis C Virus Core Antigen Monoclonal specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.
Specifications
| Hepatitis C Virus Core Antigen | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Flow Cytometry 1:100, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500 | |
| POLY | |
| Purified HCV core-GST fusion protein (genotype 1b). | |
| 0.025 mg | |
| Primary | |
| Detects hepatitis C virus (HCV) core protein from transfected human and primate cell lines. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| C7-50 | |
| Unconjugated | |
| Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 951475 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction