missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody (C7-50) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
213.00€
Specifications
| Antigen | Hepatitis C Virus Core Antigen |
|---|---|
| Clone | C7-50 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Flow Cytometry 1:100, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot |
Description
Hepatitis C Virus Core Antigen Monoclonal specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.Specifications
| Hepatitis C Virus Core Antigen | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 951475 | |
| Purified HCV core-GST fusion protein (genotype 1b). | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| C7-50 | |
| Western Blot 1 μg/mL, Flow Cytometry 1:100, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
| POLY | |
| IgG1 | |
| Immunogen affinity purified | |
| Detects hepatitis C virus (HCV) core protein from transfected human and primate cell lines. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title