missing translation for 'onlineSavingsMsg'
Learn More

Heptanal, TRC

Product Code. 30134593
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134593 25 mL 25mL
30133950 100 mL 100mL
30007819 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134593 Supplier Toronto Research Chemicals Supplier No. TRCH28114025ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-71-7
Molecular Formula C7 H14 O
Molecular Weight (g/mol) 114.19
Synonym Heptanal, 1-Heptaldehyde, Enanthal, Enanthaldehyde, Enanthic aldehyde, Enanthole, Heptaldehyde, Heptanaldehyde, Heptyl aldehyde, NSC 2190, Oenanthal, Oenanthaldehyde, Oenanthic aldehyde, Oenanthol, n-Heptaldehyde, n-Heptanal, n-Heptylaldehyde
IUPAC Name heptanal
SMILES CCCCCCC=O

Specifications

Quantity 25 mL
InChI Formula InChI=1S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3
Formula Weight 114.1045
Chemical Name or Material 1-Heptanal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.