missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexachloroethane-13C1, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 g
missing translation for 'unitSize'
0.5g
Specifications
Specifications
| Chemical Name or Material | Hexachloroethane 13 C |
| CAS | 93952-15-9 |
| Molecular Formula | 13 C C Cl6 |
| Quantity | 0.5 g |
| Synonym | Ethane-13 C, hexachloro- (9 CI), Hexachloroethane-13C1, Hexachloroethane-13 C |
| InChI Formula | InChI=1 S/C2Cl6/c3-1(4,5)2(6,7)8/i1+1 |
| SMILES | Cl[13 C](Cl)(Cl)C(Cl)(Cl)Cl |
| IUPAC Name | 1,1,1,2,2,2-hexachloro(113 C)ethane |
| Molecular Weight (g/mol) | 237.7321 |
| Formula Weight | 234.8165 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction