missing translation for 'onlineSavingsMsg'
Learn More

Hexachlorophene, TRC

Product Code. 30408393
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408393 5 g
30395064 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30408393 Supplier Toronto Research Chemicals Supplier No. TRCH2908105G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-30-4
Molecular Formula C13 H6 Cl6 O2
Molecular Weight (g/mol) 406.9
Synonym 2,2',3,3',5,5'-Hexachloro-6,6'-dihydroxydiphenylmethane, 2,2'-Dihydroxy-3,3',5,5',6,6'-hexachlorodiphenylmethane, 2,2'-Dihydroxy-3,5,6,3',5',6'-hexachlorodiphenylmethane, 2,2'-Methylenebis[3,4,6-trichlorophenol], AT 7, Acigena, Almederm, B 32, Bilevon, Bis(2-hydroxy-3,5,6-trichlorophenyl)methane, Bis(3,5,6-trichloro-2-hydroxyphenyl)methane, Cotofilm, Dermadex, Distodin, Exofene, Fesia-sin, Fostril, G 11, G 11 (bactericide), Gamophen, Gamophene, Germa-Medica, Hexabalm, Hexachlorofen, Hexachlorophen, Hexachlorophene, Hexafen, Hexophene, Hexosan, NSC 49115, NSC 9887, Nabac, Neosept V, Ritosept, Septisol, Septofen, Soy-Dome, Steral, Steraskin, Surgi-Cen, Surofene, Tersaseptic, pHisoHex, Phenol, 2,2'-methylenebis[3,4,6-trichloro-
IUPAC Name 3,4,6-trichloro-2-[(2,3,5-trichloro-6-hydroxyphenyl)methyl]phenol
SMILES Oc1c(Cl)cc(Cl)c(Cl)c1Cc2c(O)c(Cl)cc(Cl)c2Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H6Cl6O2/c14-6-2-8(16)12(20)4(10(6)18)1-5-11(19)7(15)3-9(17)13(5)21/h2-3,20-21H,1H2
Formula Weight 403.8499
Chemical Name or Material Hexachlorophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.