missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic-d11 Acid, TRC

Product Code. 30179278
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
100 mg
Unit Size:
100mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179278 2.5 mg 2.5mg
30179329 100 mg 100mg
30179530 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179278 Supplier Toronto Research Chemicals Supplier No. TRCH2650652.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95348-44-0
Molecular Formula C6 D11 H O2
Molecular Weight (g/mol) 127.23
Synonym Hexanoic-d11 acid (7CI, 9CI, ACI), Hexanoic acid-d11, Hexanoic-D11 acid, Caproic-d11 acid
IUPAC Name 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoic acid
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3,2D2,3D2,4D2,5D2
Formula Weight 127.1528
Chemical Name or Material Hexanoic acid-d11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.