missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-45266-0.025ml
This item is not returnable.
View return policy
Description
Specifically detects Histamine in All samples, and it is validated for Dot Blot, Immunohistochemistry (Paraffin)
Histamine Polyclonal specifically detects Histamine in Human, All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot.
Specifications
| Histamine | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:100 - 1:200, Dot Blot 1:14000 - 1:28000 | |
| 2 (4 imidazolyl)ethylamine, 4 (2 aminoethyl) 1 3 diazole | |
| Histamine conjugated to succinylated KLH. | |
| 0.025 mL | |
| Primary | |
| All species | |
| IgG |
| Immunohistochemistry (Paraffin), Dot Blot | |
| Unconjugated | |
| 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| The product specifically stains histamine-containing cells in paraformaldehyde/carbodiimide-fixed or paraformaldehyde-fixed, paraffin-embedded sections of rat stomach (endocrine cells and mast cells). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction