missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68393-1-IG-20UL
This item is not returnable.
View return policy
Description
The Histone H2B antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Histone H2B. This antibody recognizes human, mouse, rat, chicken, zebrafish antigen. The Histone H2B antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.Specifications
| Histone H2B | |
| Monoclonal | |
| 1000 μg/mL | |
| GL105; H2B; H2B.1; H2B/q; H2BFQ; MGC119802; MGC119804; MGC129733; MGC129734 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Chicken, Zebrafish | |
| 14-17 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 1E12G3 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 8349 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction