missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | Histone H2B |
|---|---|
| Clone | 1E12G3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Monoclonal |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
30235630
|
Proteintech
68393-1-IG-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235539
|
Proteintech
68393-1-IG-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
The Histone H2B antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Histone H2B. This antibody recognizes human, mouse, rat, chicken, zebrafish antigen. The Histone H2B antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.Specifikationer
| Histone H2B | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 8349 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1E12G3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Chicken, Zebrafish | |
| GL105; H2B; H2B.1; H2B/q; H2BFQ; MGC119802; MGC119804; MGC129733; MGC129734 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel