missing translation for 'onlineSavingsMsg'
Learn More

Histone H2B type 2E Antibody, Novus Biologicals™

Product Code. p-200059726 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18620637 100 μg 100µL
18698026 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18620637 Supplier Novus Biologicals Supplier No. NBP299782100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histone H2B type 2E Polyclonal antibody specifically detects Histone H2B type 2E in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2B type 2E
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:10000-1:20000
Formulation 0.2 um filtered solution in PBS
Gene Alias GL105, H2B, H2B Histone Family, Member Q, H2B.1, Histone 2, H2be, Histone Cluster 2 H2B Family Member E, Histone Cluster 2, H2be, Histone H2B Type 2-E, Histone H2B.Q, Histone H2B-GL105
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Histone H2B type 2E (Uniprot#: Q16778; Met1-Lys126)
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8349
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.