missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (6B8-2F2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59338-25ug
This item is not returnable.
View return policy
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| HO-1/HMOX1/HSP32 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.1% Sodium Azide | |
| HMOX1 | |
| His-tagged Rat HO-1 | |
| 25 μg | |
| Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
| 3162 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 6B8-2F2 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects 32kDa. Does not cross-react with HO-2. | |
| Store at -20°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction