missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (6B8-2F2), Novus Biologicals™
Mouse Monoclonal Antibody
174.00€ - 567.00€
Specifications
| Antigen | HO-1/HMOX1/HSP32 |
|---|---|
| Clone | 6B8-2F2 |
| Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18219283
|
Novus Biologicals
NBP2-59338 |
100 μg |
567.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603389
|
Novus Biologicals
NBP2-59338-25ug |
25 μg |
174.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| HO-1/HMOX1/HSP32 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 3162 | |
| His-tagged Rat HO-1 | |
| Primary | |
| Detects 32kDa. Does not cross-react with HO-2. |
| 6B8-2F2 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| HMOX1 | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title