missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXD13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15932465
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15932465 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15932465 Supplier Invitrogen™ Supplier No. PA575937

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 96% (by SDS-PAGE).

HOXD13 (homeobox protein D13) belongs to a class of transcription factors called homeobox proteins. In vertebrates, homeobox are found in clusters called A, B, C, and D on four separate chromosomes. Expression of homeobox proteins is spatially and temporally regulated during embryonic development. The gene encoding HOXD13 is part of the D cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. Mutations in the HOXD13 gene cause synpolydactyly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD13
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXD13
Gene Accession No. P35453, P70217
Gene Alias BDE; BDSD; homeo box 4I; homeo box D13; homeobox D13; homeobox protein Hox-4.8; Homeobox protein Hox-4I; homeobox protein Hox-D13; Hox-4.8; HOX4I; Hoxd13; SPD; SPD1; spdh
Gene Symbols HOXD13
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 173-218 of Human HOXD13.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15433, 288154, 3239
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.