missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27108
This item is not returnable.
View return policy
Description
HOXD8 Polyclonal specifically detects HOXD8 in Human, Mouse, Primate samples. It is validated for Western Blot.
Specifications
| HOXD8 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| homeo box 4E, homeo box D8, homeobox D8, homeobox protein 5.4, Homeobox protein Hox-4E, Homeobox protein Hox-5.4, homeobox protein Hox-D8, HOX4, HOX4EHox-4.5, HOX5.4 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3234 | |
| Human, Mouse, Primate | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 5-10ug/ml | |
| P13378 | |
| HOXD8 | |
| A portion of amino acids 250-290 of human HOXD8 was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in cow, 74% homologous in opossum and 66% in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction