missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 351.00€
Specifications
| Antigen | HOXD8 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 5-10ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18193265
|
Novus Biologicals
NBP2-27108SS |
0.025 mg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18169114
|
Novus Biologicals
NBP2-27108 |
0.1 mg |
351.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXD8 Polyclonal specifically detects HOXD8 in Human, Mouse, Primate samples. It is validated for Western Blot.Specifications
| HOXD8 | |
| Western Blot 5-10ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| homeo box 4E, homeo box D8, homeobox D8, homeobox protein 5.4, Homeobox protein Hox-4E, Homeobox protein Hox-5.4, homeobox protein Hox-D8, HOX4, HOX4EHox-4.5, HOX5.4 | |
| HOXD8 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in cow, 74% homologous in opossum and 66% in mouse. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Primate | |
| P13378 | |
| 3234 | |
| A portion of amino acids 250-290 of human HOXD8 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title