missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HPV18 E6 Monoclonal specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
Specifications
| Antigen | HPV18 E6 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | C1N1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Alias | E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 |
| Host Species | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?