missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Defensin 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8198-SP
This item is not returnable.
View return policy
Description
alpha-Defensin 1 Monoclonal specifically detects alpha-Defensin 1 in Human samples. It is validated for ELISA.
Specifications
| alpha-Defensin1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| alpha 2, alphaDefensin 1, DEF1, DEF1DEFA1B, DEFA1, DEFA2, defensin, alpha 1, myeloid-related sequence, defensin, alpha 1HP-1, HNP-1, HNP-1HP1, HP1, MRS, MRSMGC138393, myeloid-related sequence, neutrophil defensin 1 | |
| E. coli-derived recombinant human alpha-Defensin 1 Ala65-Cys94 Accession # P59665 | |
| 25 μg | |
| 1667 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 867919R | |
| ELISA | |
| P59665 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human alpha-Defensin 1 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction