missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Defensin 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
245.00€ - 452.00€
Specifications
| Antigen | alpha-Defensin1 |
|---|---|
| Clone | 867919R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
18070702
![]() |
R&D Systems
MAB8198-SP |
25 μg |
245.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18059051
|
R&D Systems
MAB8198-100 |
100 μg |
452.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
alpha-Defensin 1 Monoclonal specifically detects alpha-Defensin 1 in Human samples. It is validated for ELISA.Spezifikation
| alpha-Defensin1 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P59665 | |
| 1667 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 867919R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| alpha 2, alphaDefensin 1, DEF1, DEF1DEFA1B, DEFA1, DEFA2, defensin, alpha 1, myeloid-related sequence, defensin, alpha 1HP-1, HNP-1, HNP-1HP1, HP1, MRS, MRSMGC138393, myeloid-related sequence, neutrophil defensin 1 | |
| E. coli-derived recombinant human alpha-Defensin 1 Ala65-Cys94 Accession # P59665 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human alpha-Defensin 1 in direct ELISAs. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts