missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AMICA/JAML Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3449
This item is not returnable.
View return policy
Description
AMICA/JAML Monoclonal specifically detects AMICA/JAML in Human samples. It is validated for Western Blot.
Specifications
| AMICA/JAML | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q86YT9 | |
| AMICA1 | |
| Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML Leu20-Leu275 Accession # Q86YT9 | |
| 100 μg | |
| Primary | |
| Detects human AMICA/JAML in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse AMICA is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 401910 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 120425 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction