missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AMPKβ1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11642-SP
This item is not returnable.
View return policy
Description
AMPK beta 1 Monoclonal antibody specifically detects AMPK beta 1 in Human samples. It is validated for Western Blot,Immunohistochemistry,Simple Western
Specifications
| AMPK beta 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5'-AMP-activated protein kinase beta-1 subunit, AMP-activated protein kinase beta subunit, AMPK, AMPK beta -1 chain, AMPK beta 1,5'-AMP-activated protein kinase subunit beta-1, AMPK subunit beta-1, AMPKb, HAMPKb, MGC17785, PRKAB1, protein kinase, AMP-activated, beta 1 non-catalytic subunit, protein kinase, AMP-activated, noncatalytic, beta-1 | |
| E. coli- derived recombinant human AMPKbeta1, Met1-Ile270, Accession # Q9Y478 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Simple Western | |
| 1092531 | |
| Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 0.5-25 μg/mL | |
| Q9Y478 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5564 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction