missing translation for 'onlineSavingsMsg'
Learn More

Human Atrial Natriuretic Peptide/ANP Antibody, R&D Systems™

Product Code. 18010462
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18010462 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18010462 Supplier R&D Systems Supplier No. AF3366

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Atrial Natriuretic Peptide/ANP Polyclonal specifically detects Atrial Natriuretic Peptide/ANP in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atrial Natriuretic Peptide/ANP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P01160
Gene Alias ANF, ANP, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, Cardionatrin, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, NPPA, PND, PNDcardionatrin, prepronatriodilatin
Gene Symbols NPPA
Host Species Goat
Immunogen E. coli-derived recombinant human Atrial Natriuretic Peptide/ANP Asn26-Tyr151 Accession # P01160
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4878
Test Specificity Detects human Atrial Natriuretic Peptide/ANP in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.