missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Azurocidin/CAP37/HBP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2200-SP
This item is not returnable.
View return policy
Description
Azurocidin/CAP37/HBP Polyclonal specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Azurocidin/CAP37/HBP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AZAMP, AZU, AZU1, azurocidin, azurocidin 1, CAP37, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBP, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 566 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P20160 | |
| AZU1 | |
| Mouse myeloma cell line NS0-derived recombinant human Azurocidin/CAP37/HBP Ile27-Pro250 Accession # P20160 | |
| 25 μg | |
| Primary | |
| Detects human Azurocidin/CAP37/HBP in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction