missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Azurocidin/CAP37/HBP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
209.00€ - 605.00€
Specifications
| Antigen | Azurocidin/CAP37/HBP |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765047
![]() |
R&D Systems
AF2200-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18092522
|
R&D Systems
AF2200 |
100 μg |
605.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Azurocidin/CAP37/HBP Polyclonal specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Azurocidin/CAP37/HBP | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| AZAMP, AZU, AZU1, azurocidin, azurocidin 1, CAP37, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBP, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin | |
| AZU1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P20160 | |
| 566 | |
| Mouse myeloma cell line NS0-derived recombinant human Azurocidin/CAP37/HBP Ile27-Pro250 Accession # P20160 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Azurocidin/CAP37/HBP in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title